Gene Transcripts

Gene Symbol
Met
Entrez Gene ID
17295
Gene Name
met proto-oncogene
Gene Aliases
AI838057, HGF, HGFR, Par4, c-Met
Chromosome Location
Chr. 6: 17463349 - 17573980 on Build GRCm38
UniGene ID
Mm.86844
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008591.2 NP_032617.2 3 1504
GenBank mRNA BC117969.1 - 1 1148
BC117970.1 - 1 1144
Y00671.1 - 1 1149

View Full Product Details