Gene Transcripts

Gene Symbol
Zfp354a
Entrez Gene ID
21408
Gene Name
zinc finger protein 354A
Gene Aliases
AW488485, Tcf17, Znf354a, kid1
Chromosome Location
Chr. 11: 51059244 - 51072799 on Build GRCm38
UniGene ID
Mm.4792
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009329.3 NP_033355.2 5 624
GenBank mRNA AB221586.1 - 4 428
AF184111.1 - 5 586
AK030469.1 - 5 603
BC050843.1 - 5 624 6506256
BC087540.1 - 5 760 4239789
L77247.1 - 4 488

View Full Product Details