Gene Transcripts

Gene Symbol
Ckap4
Entrez Gene ID
216197
Gene Name
cytoskeleton-associated protein 4
Gene Aliases
5630400A09Rik, CLIMP-63, P63
Chromosome Location
Chr. 10: 84526305 - 84533888 on Build GRCm38
UniGene ID
Mm.334999
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175451.1 NP_780660.1 2 898
GenBank mRNA AK030708.1 - 2 782
AK089935.1 - 1 144
AK132136.1 - 1072
BC025522.1 - 2 549
BC138318.1 - 3 920 8861338
BC138320.1 - 3 920 8861340

View Full Product Details