Gene Transcripts

Gene Symbol
Sept7
Entrez Gene ID
235072
Gene Name
septin 7
Gene Aliases
Cdc10, E430034N22
Chromosome Location
Chr. 9: 25251090 - 25308571 on Build GRCm38
UniGene ID
Mm.270259
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205367.1 NP_001192296.1 2 307
NM_009859.4 NP_033989.2 3 310
GenBank mRNA AK088997.1 - 3 97
AK141572.1 - 3 102
AK144127.1 - 3 99
AK147986.1 - 3 102
AK220447.1 - 2 80
BC058587.1 - 2 111 6529169

View Full Product Details