Gene Transcripts

Gene Symbol
Gkn3
Entrez Gene ID
68888
Gene Name
gastrokine 3
Gene Aliases
1190003M12Rik
Chromosome Location
Chr. 6: 87383319 - 87388935 on Build GRCm38
UniGene ID
Mm.46387
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026860.1 NP_081136.1 5 471
GenBank mRNA AK004474.1 - 5 391
BC116955.1 - 5 471 40126274
FN600638.1 - 5 390
GU220566.1 - 5 390

View Full Product Details