Gene Transcripts

Gene Symbol
Wdr13
Entrez Gene ID
73447
Gene Name
WD repeat domain 13
Gene Aliases
1700060B08Rik, 5730411P10Rik, DXHXS7467e, W51679, mMg21
Chromosome Location
Chr. X: 8123301 - 8132858 on Build GRCm38
UniGene ID
Mm.284162
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290783.1 NP_001277712.1 7 1555
NM_026137.5 NP_080413.2 7 1589
NM_028542.1 NP_082818.1 7 1841
GenBank mRNA AF220146.1 - 6 1111
AF353243.1 - 7 1235
AF513713.1 - 7 1058
AK006837.1 - 6 1077
AK017549.1 - 7 1171
BC057366.1 - 7 1555
BC080853.1 - 7 1504
BC108332.1 - 7 1345 6493226
BC120553.1 - 7 1092 40129476
BC125287.1 - 7 1091 40129802

View Full Product Details