Gene Transcripts

Gene Symbol
Ppp1r21
Entrez Gene ID
73825
Gene Name
protein phosphatase 1, regulatory subunit 21
Gene Aliases
1110018J12Rik, AI426045, AW550781, Ccdc128, Klraq1
Chromosome Location
Chr. 17: 88530124 - 88588367 on Build GRCm38
UniGene ID
Mm.271817
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028658.4 NP_082934.3 12 1326
GenBank mRNA AK028185.1 - 12 1190
AK155094.1 - 12 1158
AK170254.1 - 12 1214
BC137673.1 - 12 1266 8860692

View Full Product Details