Gene Transcripts

Gene Symbol
Spata17
Entrez Gene ID
74717
Gene Name
spermatogenesis associated 17
Gene Aliases
1700065F16Rik, 4930504I07Rik, 4930513F16Rik, MSRG-11
Chromosome Location
Chr. 1: 187009614 - 187215462 on Build GRCm38
UniGene ID
Mm.250430
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028848.3 NP_083124.1 5 524
NM_029067.1 NP_083343.1 5 524
GenBank mRNA AY747687.1 - 5 499
BC089520.1 - 5 499 6774024

View Full Product Details