Gene Transcripts

Gene Symbol
Pcf11
Entrez Gene ID
74737
Gene Name
cleavage and polyadenylation factor subunit homolog (S. cerevisiae)
Gene Aliases
2500001H09Rik, 5730417B17Rik, C77803
Chromosome Location
Chr. 7: 92643458 - 92670087 on Build GRCm38
UniGene ID
Mm.288645
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029078.3 NP_083354.3 3 700
GenBank mRNA AK161260.1 - 3 724
AK161614.1 - 3 690
AK173035.1 - 3 672
BC150781.1 - 3 536 9087692

View Full Product Details