Gene Transcripts

Gene Symbol
Ccdc150
Entrez Gene ID
78016
Gene Name
coiled-coil domain containing 150
Gene Aliases
4930511H11Rik
Chromosome Location
Chr. 1: 54250661 - 54368727 on Build GRCm38
UniGene ID
Mm.67455
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030025.2 NP_084301.2 26, 27 3258
GenBank mRNA AK029994.1 - 26, 27 3258

View Full Product Details