Gene Transcripts

Gene Symbol
Olfr594
Entrez Gene ID
258246
Gene Name
olfactory receptor 594
Gene Aliases
MOR32-10
Chromosome Location
Chr. 7: 103219720 - 103220655 on Build GRCm38
UniGene ID
Mm.377390
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207143.1 NP_997026.1 1 785
GenBank mRNA BC141016.1 - 1 862

View Full Product Details