Gene Transcripts

Gene Symbol
Hexb
Entrez Gene ID
15212
Gene Name
hexosaminidase B
Gene Aliases
Chromosome Location
Chr. 13: 97176331 - 97198357 on Build GRCm38
UniGene ID
Mm.27816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010422.2 NP_034552.1 8, 9 1106
GenBank mRNA AK151149.1 - 8, 9 1043
AK159134.1 - 8, 9 1059
AK159178.1 - 8, 9 1080
AK160560.1 - 8, 9 1075
AK168825.1 - 8, 9 1080
AK194154.1 - 1, 2 88
AK212361.1 - 1, 2 88
U07633.1 - 8, 9 1078
Y00964.1 - 8, 9 1019

View Full Product Details