Gene Transcripts

Gene Symbol
Chdh
Entrez Gene ID
218865
Gene Name
choline dehydrogenase
Gene Aliases
AU041472, D630034H06Rik
Chromosome Location
Chr. 14: 30009000 - 30040466 on Build GRCm38
UniGene ID
Mm.259916
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136240.1 NP_001129712.1 8 1966
NM_172264.2 NP_758468.2 8 1764
NM_175343.5 NP_780552.1 8 1939
GenBank mRNA AK030900.1 - 8 1940
AK042454.1 - 1 262
AK052715.1 - 8 1947
AK087570.1 - 8 1875
AK164042.1 - 8 1802
BC039186.1 - 8 1764 3979709

View Full Product Details