Gene Transcripts

Gene Symbol
Crygc
Entrez Gene ID
12966
Gene Name
crystallin, gamma C
Gene Aliases
Cryg-5
Chromosome Location
Chr. 1: 65071525 - 65073532 on Build GRCm38
UniGene ID
Mm.491115
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082573.2 NP_001076042.1 3 468
NM_007775.2 NP_031801.1 3 465
GenBank mRNA BC056454.1 - 2 444
CJ066821.1 - 3 466
CK627566.1 - 1 63
EF426301.1 - 3 454
EF426302.1 - 3 457
M64544.1 - 2 417

View Full Product Details