Gene Transcripts

Gene Symbol
Gstm1
Entrez Gene ID
14862
Gene Name
glutathione S-transferase, mu 1
Gene Aliases
Gstb-1, Gstb1
Chromosome Location
Chr. 3: 108012250 - 108017973 on Build GRCm38
UniGene ID
Mm.37199
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010358.5 NP_034488.1 1, 2 222
GenBank mRNA BC003822.1 - 1, 2 45
BC046758.1 - 1, 2 75 5693552
BC091763.1 - 1, 2 55 5102093
J03952.1 - 1, 2 36
J04632.1 - 1, 2 223

View Full Product Details