Gene Transcripts

Gene Symbol
Odc1
Entrez Gene ID
18263
Gene Name
ornithine decarboxylase, structural 1
Gene Aliases
ODC
Chromosome Location
Chr. 12: 17544873 - 17551502 on Build GRCm38
UniGene ID
Mm.34102
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013614.2 NP_038642.2 11 1520
GenBank mRNA AK077679.1 - 11 1423
AK139610.1 - 11 1422
AK150283.1 - 11 1422
AK150343.1 - 11 1421
AK152525.1 - 11 1420
AK166093.1 - 11 1427
AK168219.1 - 11 1421
AK169214.1 - 11 1421
AK172568.1 - 11 1423
AK190288.1 - 2 208
AK202063.1 - 209
AK202843.1 - 2 208
BC033264.1 - 11 1411 2645291
BC059826.1 - 11 1401 6808137
BC083122.1 - 11 1408 6329274
EU684749.1 - 9 1110
M12330.1 - 6 579
M12331.1 - 2 155
M20617.2 - 9 1126
S64539.1 - 11 1420

View Full Product Details