Gene Transcripts

Gene Symbol
Lrtm2
Entrez Gene ID
211187
Gene Name
leucine-rich repeats and transmembrane domains 2
Gene Aliases
A230084J22, AI841794
Chromosome Location
Chr. 6: 119315133 - 119332270 on Build GRCm38
UniGene ID
Mm.121498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172207.1 NP_001165678.1 4 631
NM_172492.3 NP_766080.1 4 802
GenBank mRNA AK039006.1 - 4 636
AK045494.1 - 4 716
BC058944.1 - 3 1192 6827376

View Full Product Details