Gene Transcripts

Gene Symbol
Mmgt2
Entrez Gene ID
216829
Gene Name
membrane magnesium transporter 2
Gene Aliases
AI852046
Chromosome Location
Chr. 11: 62648632 - 62666359 on Build GRCm38
UniGene ID
Mm.295306
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175002.2 NP_778167.1 2 522
GenBank mRNA AK004099.1 - 2 359
AK131977.1 - 2 412
BC025076.1 - 1 293 4459181
BC096583.1 - 2 466 5025927
BI134823.1 - 1 349
EU069462.1 - 1 180

View Full Product Details