Gene Transcripts

Gene Symbol
Rbm27
Entrez Gene ID
225432
Gene Name
RNA binding motif protein 27
Gene Aliases
A730010I06, AI043120, Psc1, mKIAA1311
Chromosome Location
Chr. 18: 42275353 - 42341542 on Build GRCm38
UniGene ID
Mm.329400
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172626.2 NP_766214.2 8 1415
GenBank mRNA AK042613.1 - 3 648
AK129330.1 - 2 259
AY461716.1 - 8 1390
BC054080.1 - 4 863 6390990

View Full Product Details