Gene Transcripts

Gene Symbol
Pip5kl1
Entrez Gene ID
227733
Gene Name
phosphatidylinositol-4-phosphate 5-kinase-like 1
Gene Aliases
BC028795
Chromosome Location
Chr. 2: 32574788 - 32583780 on Build GRCm38
UniGene ID
Mm.78923
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198191.2 NP_937834.1 3 298
GenBank mRNA AK164998.1 - 3 401
AY376879.1 - 3 337
BC094346.1 - 3 399 6411828
BC117018.1 - 3 290 40126337
BC119037.1 - 3 290 8733786

View Full Product Details