Gene Transcripts

Gene Symbol
Rft1
Entrez Gene ID
328370
Gene Name
RFT1 homolog (S. cerevisiae)
Gene Aliases
D930025H04
Chromosome Location
Chr. 14: 30654341 - 30691317 on Build GRCm38
UniGene ID
Mm.259657
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177815.3 NP_808483.2 10 1114
GenBank mRNA BC082539.1 - 10 1089
BC094336.1 - 10 1082
BC141275.1 - 10 1108 9055887

View Full Product Details