Gene Transcripts

Gene Symbol
Dmrtb1
Entrez Gene ID
56296
Gene Name
DMRT-like family B with proline-rich C-terminal, 1
Gene Aliases
Dmrt6, Prp13
Chromosome Location
Chr. 4: 107676288 - 107684243 on Build GRCm38
UniGene ID
Mm.141324
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019872.1 NP_063925.1 1 498
GenBank mRNA AF542048.1 - 1 498

View Full Product Details