Gene Transcripts

Gene Symbol
B230219D22Rik
Entrez Gene ID
78521
Gene Name
RIKEN cDNA B230219D22 gene
Gene Aliases
AA119856, D530037I19Rik
Chromosome Location
Chr. 13: 55693124 - 55703500 on Build GRCm38
UniGene ID
Mm.28449
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181278.2 NP_851795.1 2 248
GenBank mRNA AJ492504.1 - 1 39
AK088693.1 - 2 265
BC051430.1 - 2 248 6530869

View Full Product Details