Gene Transcripts

Gene Symbol
Hey2
Entrez Gene ID
155430
Gene Name
hes-related family bHLH transcription factor with YRPW motif 2
Gene Aliases
Herp1
Chromosome Location
Chr. 1: 29191170 - 29201257 on Build Rnor_6.0
UniGene ID
Rn.58672
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130417.1 NP_569101.1 5 1037
GenBank mRNA AY059382.1 - 1 510

View Full Product Details