Gene Transcripts

Gene Symbol
Mrgprx3
Entrez Gene ID
252960
Gene Name
MAS-related GPR, member X3
Gene Aliases
Mgrg1, MrgA, Mrga10, Mrgpra
Chromosome Location
Chr. 1: 103298172 - 103323476 on Build Rnor_6.0
UniGene ID
Rn.81069
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145787.2 NP_665730.2 2 628
GenBank mRNA AJ311952.1 - 1 574
BC089774.1 - 2 628 7321342

View Full Product Details