Gene Transcripts

Gene Symbol
Sstr4
Entrez Gene ID
25555
Gene Name
somatostatin receptor 4
Gene Aliases
Smstr4
Chromosome Location
Chr. 3: 142739781 - 142741644 on Build Rnor_6.0
UniGene ID
Rn.9936
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013036.2 NP_037168.1 1 193
GenBank mRNA U04738.1 - 1 193

View Full Product Details