Gene Transcripts

Gene Symbol
Tsen34
Entrez Gene ID
292534
Gene Name
tRNA splicing endonuclease 34 homolog (S. cerevisiae)
Gene Aliases
LENG5, SEN34
Chromosome Location
Chr. 1: 64023924 - 64031755 on Build Rnor_6.0
UniGene ID
Rn.1591
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006968.1 NP_001006969.1 539
XM_006228017.2 XP_006228079.1 973
XM_006228018.2 XP_006228080.1 547
XM_008758752.1 XP_008756974.1 1152
GenBank mRNA BC083767.1 - 539

Gene Symbol
LOC100909510
Entrez Gene ID
100909510
Gene Name
tRNA-splicing endonuclease subunit Sen34-like
Gene Aliases
Chromosome Location
Chr. 1: 64092224 - 64100055 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748716.3 XP_003748764.1 3 909
XM_003748717.3 XP_003748765.1 3 547
XM_006228075.2 XP_006228137.1 3 973
XM_008758839.1 XP_008757061.1 2 1152

View Full Product Details