Gene Transcripts

Gene Symbol
Kcnq4
Entrez Gene ID
298496
Gene Name
potassium channel, voltage-gated KQT-like subfamily Q, member 4
Gene Aliases
Chromosome Location
Chr. 5: 139625783 - 139677300 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764109.1 XP_008762331.1 11, 12 1623
XM_008764110.1 XP_008762332.1 10, 11 1461
XM_008764111.1 XP_008762333.1 11, 12 1348
GenBank mRNA AF249748.1 - 2, 3 313

View Full Product Details