Gene Transcripts

Gene Symbol
Slc35a3
Entrez Gene ID
310808
Gene Name
solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3
Gene Aliases
Chromosome Location
Chr. 2: 219702681 - 219741886 on Build Rnor_6.0
UniGene ID
Rn.162630
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012082.1 NP_001012082.1 2 206
XM_008761461.1 XP_008759683.1 2 474
GenBank mRNA BC079371.1 - 2 206

View Full Product Details