Gene Transcripts

Gene Symbol
Olr1283
Entrez Gene ID
315572
Gene Name
olfactory receptor 1283
Gene Aliases
tpcr21
Chromosome Location
Chr. 15: 35813468 - 35814520 on Build Rnor_6.0
UniGene ID
Rn.120679
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000518.1 NP_001000518.1 1 574
GenBank mRNA X89704.1 - 1 191

View Full Product Details