Gene Transcripts

Gene Symbol
Btbd7
Entrez Gene ID
362772
Gene Name
BTB (POZ) domain containing 7
Gene Aliases
Chromosome Location
Chr. 6: 126658398 - 126747883 on Build Rnor_6.0
UniGene ID
Rn.21901
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108720.1 NP_001102190.1 7 2030
XM_008764818.1 XP_008763040.1 7 2065
XM_008764819.1 XP_008763041.1 8 2207

View Full Product Details