Gene Transcripts

Gene Symbol
Phka1
Entrez Gene ID
64561
Gene Name
phosphorylase kinase, alpha 1
Gene Aliases
Pcyt1b
Chromosome Location
Chr. X: 72377020 - 72515387 on Build Rnor_6.0
UniGene ID
Rn.218907
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022626.1 NP_072148.1 19 2090
XM_006257101.2 XP_006257163.1 19 2417
GenBank mRNA AF197561.1 - 19 2090

View Full Product Details