Gene Transcripts

Gene Symbol
Gnrhr
Entrez Gene ID
81668
Gene Name
gonadotropin releasing hormone receptor
Gene Aliases
GH1, Lhrhr
Chromosome Location
Chr. 14: 23480462 - 23498450 on Build Rnor_6.0
UniGene ID
Rn.112595
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031038.3 NP_112300.2 2 747
GenBank mRNA L07646.1 - 2 733
X68980.1 - 2 697
X76635.1 - 2 686

View Full Product Details