Gene Transcripts

Gene Symbol
Foxg1
Entrez Gene ID
24370
Gene Name
forkhead box G1
Gene Aliases
BF1A, Fkhl1, RATBF1A
Chromosome Location
Chr. 6: 69971227 - 69974037 on Build Rnor_6.0
UniGene ID
Rn.9864
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012560.2 NP_036692.1 1 1442
GenBank mRNA M87634.1 - 1 1443

View Full Product Details