Gene Transcripts

Gene Symbol
Olr709
Entrez Gene ID
405954
Gene Name
olfactory receptor 709
Gene Aliases
Chromosome Location
Chr. 3: 78525078 - 78526022 on Build Rnor_6.0
UniGene ID
Rn.142806
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001066.1 NP_001001066.1 1 464

View Full Product Details