Gene Transcripts

Gene Symbol
Dhx33
Entrez Gene ID
287464
Gene Name
DEAH (Asp-Glu-Ala-His) box polypeptide 33
Gene Aliases
Chromosome Location
Chr. 10: 57622022 - 57638485 on Build Rnor_6.0
UniGene ID
Rn.198877
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105802.1 NP_001099272.1 12 1916

View Full Product Details