Gene Transcripts

Gene Symbol
Gpr97
Entrez Gene ID
291854
Gene Name
G protein-coupled receptor 97
Gene Aliases
Chromosome Location
Chr. 19: 10392001 - 10422604 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006255126.2 XP_006255188.1 11 3304
XM_006255127.2 XP_006255189.1 10 3157

View Full Product Details