Gene Transcripts

Gene Symbol
Agtrap
Entrez Gene ID
298646
Gene Name
angiotensin II receptor-associated protein
Gene Aliases
Chromosome Location
Chr. 5: 164888069 - 164898387 on Build Rnor_6.0
UniGene ID
Rn.8471
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007654.1 NP_001007655.1 1, 2 65
GenBank mRNA BC082019.1 - 1, 2 65

View Full Product Details