Gene Transcripts

Gene Symbol
Rnf151
Entrez Gene ID
302977
Gene Name
ring finger protein 151
Gene Aliases
Chromosome Location
Chr. 10: 14083538 - 14088025 on Build Rnor_6.0
UniGene ID
Rn.89276
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106987.1 NP_001100457.1 4 351
XM_006245952.2 XP_006246014.1 5 527

View Full Product Details