Gene Transcripts

Gene Symbol
Ccno
Entrez Gene ID
499528
Gene Name
cyclin O
Gene Aliases
RGD1565217
Chromosome Location
Chr. 2: 44855561 - 44860317 on Build Rnor_6.0
UniGene ID
Rn.61269
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109175.1 NP_001102645.1 2 1649
XM_006231947.2 XP_006232009.1 3 2781

View Full Product Details