Gene Transcripts

Gene Symbol
Rab27a
Entrez Gene ID
50645
Gene Name
RAB27A, member RAS oncogene family
Gene Aliases
ram
Chromosome Location
Chr. 8: 79722334 - 79776222 on Build Rnor_6.0
UniGene ID
Rn.37360
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017317.2 NP_059013.1 6 586
GenBank mRNA BC098667.1 - 6 586
D17352.1 - 5 522

View Full Product Details