Gene Transcripts

Gene Symbol
Cldn5
Entrez Gene ID
65131
Gene Name
claudin 5
Gene Aliases
Chromosome Location
Chr. 11: 86356292 - 86357718 on Build Rnor_6.0
UniGene ID
Rn.17420
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031701.2 NP_113889.1 1 419
GenBank mRNA AF241260.1 - 1 250
BC082073.1 - 1 417 7133096
CK479973.1 - 1 419
FQ213419.1 - 1 432

View Full Product Details