Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_174899.4 | NP_777559.3 |
XM_005246317.1 | XP_005246374.1 | |
GenBank mRNA | AK094911.1 | - |
AK123085.1 | - | |
AK289655.1 | - | |
AK309702.1 | - | |
BC017869.1 | - | |
BC022538.1 | - | |
BC033935.1 | - | |
BF674223.1 | - | |
BM997950.1 | - | |
ENST00000283946.0 | - |