Gene Transcripts

Gene Symbol
MIR6506
Entrez Gene ID
102465252
Gene Name
microRNA 6506
Gene Aliases
hsa-mir-6506
Chromosome Location
Chr.16: 15611030 - 15611095 on Build GRCh38
Interrogated Sequence Translated Protein
RefSeq NR_106761.1

View Full Product Details