Gene Transcripts

Gene Symbol
SLC1A2
Entrez Gene ID
6506
Gene Name
solute carrier family 1 member 2
Gene Aliases
EAAT2, GLT-1, HBGT
Chromosome Location
Chr.11: 35251205 - 35420063 on Build GRCh38
Uni Gene
Hs.502338
Interrogated Sequence Translated Protein
RefSeq NM_001195728.2 NP_001182657.1
NM_001252652.1 NP_001239581.1
NM_004171.3 NP_004162.2
XM_005253067.1 XP_005253124.1
XM_011520285.2 XP_011518587.1
XM_017018136.1 XP_016873625.1
XM_017018137.1 XP_016873626.1
XM_017018138.1 XP_016873627.1
XM_017018139.1 XP_016873628.1
GenBank mRNA AB209444.1 -
AF131756.1 -
AK054855.1 -
AK057674.1 -
AK090810.1 -
AK122843.1 -
AK298769.1 -
AL157452.1 -
AY066021.1 -
BC031313.2 -
BC047052.1 -
BC132768.1 -
BX509751.1 -
BX538155.1 -
BX648750.1 -
CR626999.1 -
CR627009.1 -
CR627022.1 -
CR749842.1 -
D85884.1 -
DA158200.1 -
DA215039.1 -
DB465494.2 -
ENST00000278379.0 -
KP966086.1 -
S83374.1 -
S83375.1 -
U01824.1 -
U03505.1 -
Z32517.1 -

View Full Product Details