Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001195728.2 | NP_001182657.1 |
NM_001252652.1 | NP_001239581.1 | |
NM_004171.3 | NP_004162.2 | |
XM_005253067.1 | XP_005253124.1 | |
XM_011520285.2 | XP_011518587.1 | |
XM_017018136.1 | XP_016873625.1 | |
XM_017018137.1 | XP_016873626.1 | |
XM_017018138.1 | XP_016873627.1 | |
XM_017018139.1 | XP_016873628.1 | |
GenBank mRNA | AB209444.1 | - |
AF131756.1 | - | |
AK054855.1 | - | |
AK057674.1 | - | |
AK090810.1 | - | |
AK122843.1 | - | |
AK298769.1 | - | |
AL157452.1 | - | |
AY066021.1 | - | |
BC031313.2 | - | |
BC047052.1 | - | |
BC132768.1 | - | |
BX509751.1 | - | |
BX538155.1 | - | |
BX648750.1 | - | |
CR626999.1 | - | |
CR627009.1 | - | |
CR627022.1 | - | |
CR749842.1 | - | |
D85884.1 | - | |
DA158200.1 | - | |
DA215039.1 | - | |
DB465494.2 | - | |
ENST00000278379.0 | - | |
KP966086.1 | - | |
S83374.1 | - | |
S83375.1 | - | |
U01824.1 | - | |
U03505.1 | - | |
Z32517.1 | - |