Gene Transcripts

Gene Symbol
FBXO36
Entrez Gene ID
130888
Gene Name
F-box protein 36
Gene Aliases
Fbx36
Chromosome Location
Chr.2: 229922491 - 230013109 on Build GRCh38
Uni Gene
Hs.140666
Interrogated Sequence Translated Protein
RefSeq NM_174899.4 NP_777559.3
XM_005246317.1 XP_005246374.1
GenBank mRNA AK094911.1 -
AK123085.1 -
AK289655.1 -
AK309702.1 -
BC017869.1 -
BC022538.1 -
BC033935.1 -
BF674223.1 -
BM997950.1 -
ENST00000283946.0 -

View Full Product Details