Gene Transcripts

Gene Symbol
Emx2os
Entrez Gene ID
329078
Gene Name
Emx2 opposite strand/antisense transcript (non-protein coding)
Gene Aliases
Chromosome Location
Chr. 19: 59425104 - 59458635 on Build GRCm38
UniGene ID
Mm.80382
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002863.2
3 810
GenBank mRNA AK033011.1 -
3 459
AY117414.1 - LIT1111
EMX2OS
3 809

View Full Product Details