Gene Transcripts

Gene Symbol
Snora78
Entrez Gene ID
100306952
Gene Name
small nucleolar RNA, H/ACA box 7
Gene Aliases
ACA64
Chromosome Location
Chr. 17: 24719676 - 24719832 on Build GRCm38
UniGene ID
Mm.484806
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028515.1 1 34


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AV568172.1 - 1 90 Chr. 17 - 24719531 - 24719888 [-]

View Full Product Details