Gene Transcripts
- Gene Symbol
- 2700099C18Rik
- Entrez Gene ID
- 77022
- Gene Name
- NDC80 homolog, kinetochore complex component pseudogene
- Gene Aliases
- 9430041D22Rik
- Chromosome Location
- Chr. 17: 94750100 - 94775129 on Build GRCm38
- UniGene ID
- Mm.440259
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CF751226.1 |
- |
3 |
201 |
|
Chr. 17 - 94763901 - 94775132 [+] |
GenBankmRNA |
CF949594.1 |
- |
4 |
429 |
|
Chr. 17 - 94762522 - 94775052 [+] |
View Full Product Details